(E)-4-((4-Bromophenyl)amino)-4-oxobut-2-enoic acid ≥95%

Supplier: AOBChem USA

Warning

357571-5G 357571-25G
77493-368EA 167.89 USD
77493-368 77503-154
(E)-4-((4-Bromophenyl)amino)-4-oxobut-2-enoic acid ≥95%
(E)-4-((4-Bromophenyl)amino)-4-oxobut-2-enoic acid

Precaución: For research use only

Formula: C₁₀H₈BrNO₃
MW: 270.079 g/mol
Storage Temperature: Ambient
CAS Number: 843664-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR