4-(4-Bromophenyl)-2-methylbut-3-yn-2-ol ≥95%

Supplier: AOBChem USA

Warning

320032-5G 320032-25G
77492-230EA 149.35 USD
77492-230 77502-160
4-(4-Bromophenyl)-2-methylbut-3-yn-2-ol ≥95%
4-(4-Bromophenyl)-2-methylbut-3-yn-2-ol

Precaución: For research use only

Formula: C₁₁H₁₁BrO
MW: 239.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00168840
CAS Number: 76347-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR