1-Fluoro-4-(2-propen-1-yl)benzene ≥95%

Supplier: AOBChem USA

Warning

293708-5G 293708-25G
77491-368EA 84.46 USD
77491-368 77501-364
1-Fluoro-4-(2-propen-1-yl)benzene ≥95%
1-Fluoro-4-(2-propen-1-yl)benzene

Precaución: For research use only

Formula: C₉H₉F
MW: 136.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07698630
CAS Number: 1737-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR