2-Bromo-4-ethyl-6-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

289464-5G 289464-25G
77491-256EA 131.84 USD
77491-256 77501-266
2-Bromo-4-ethyl-6-methoxyphenol ≥95%
2-Bromo-4-ethyl-6-methoxyphenol

Precaución: For research use only

Formula: C₉H₁₁BrO₂
MW: 231.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414718
CAS Number: 1447718-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR