1,4-Bis(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

26460-5G 26460-25G
77490-820EA 90.64 USD
77490-820 77500-894
1,4-Bis(methoxymethoxy)benzene ≥97%
1,4-Bis(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₁₀H₁₄O₄
MW: 198.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27931250
CAS Number: 87905-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR