4-Bromo-3,5-dimethylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

23025-5G 23025-25G
77490-118EA 139.05 USD
77490-118 77500-348
4-Bromo-3,5-dimethylphenylboronic acid ≥95%
4-Bromo-3,5-dimethylphenylboronic acid

Precaución: For research use only

Formula: C₈H₁₀BBrO₂
Storage Temperature: Ambient
MDL Number: MFCD23713094
CAS Number: 1451391-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR