(3-(Benzyloxy)-4-chloro-2-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

21008-5G 21008-25G
77489-746EA 115.36 USD
77489-746 77500-046
(3-(Benzyloxy)-4-chloro-2-fluorophenyl)boronic acid ≥97%
(3-(Benzyloxy)-4-chloro-2-fluorophenyl)boronic acid

Precaución: For research use only

Formula: C₁₃H₁₁BClFO₃
Storage Temperature: Ambient
MDL Number: MFCD17015743
CAS Number: 1256346-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR