2-Fluoro-4-n-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20309-5G 20309-25G
77489-658EA 152.44 USD
77489-658 77499-978
2-Fluoro-4-n-propoxybenzaldehyde ≥97%
2-Fluoro-4-n-propoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247715
CAS Number: 883543-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR