4-Fluoro-2-methoxyphenyl acetate ≥95%

Supplier: AOBChem USA

Warning

16550-5G 16550-25G
77488-350EA 90.64 USD
77488-350 77499-012
4-Fluoro-2-methoxyphenyl acetate ≥95%
4-Fluoro-2-methoxyphenyl acetate

Precaución: For research use only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD27931837
CAS Number: 900174-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR