(2-Chloro-6-fluorophenyl)(ethyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

161559-5G 161559-25G
77488-170EA 60.77 USD
77488-170 77498-866
(2-Chloro-6-fluorophenyl)(ethyl)sulfane ≥97%
(2-Chloro-6-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₈ClFS
Storage Temperature: Ambient
MDL Number: MFCD34848748
CAS Number: 2921875-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR