2,4-Dichloro-1-isobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

157650-5G 157650-25G
77488-082EA 173.04 USD
77488-082 77498-782
2,4-Dichloro-1-isobutoxybenzene ≥97%
2,4-Dichloro-1-isobutoxybenzene

Precaución: For research use only

Formula: C₁₀H₁₂Cl₂O
MW: 219.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34846315
CAS Number: 6851-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR