1-(Methoxymethoxy)-3-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

13042-5G 13042-25G
77487-494EA 90.64 USD
77487-494 77498-284
1-(Methoxymethoxy)-3-(trifluoromethoxy)benzene ≥97%
1-(Methoxymethoxy)-3-(trifluoromethoxy)benzene

Precaución: For research use only

Formula: C₉H₉F₃O₃
MW: 222.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27929877
CAS Number: 851341-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR