1-Fluoro-2-iodo-4-methoxy-3-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

120829-5G 120829-25G
77487-190EA 145.23 USD
77487-190 77498-002
1-Fluoro-2-iodo-4-methoxy-3-(methoxymethoxy)benzene ≥97%
1-Fluoro-2-iodo-4-methoxy-3-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₉H₁₀FIO₃
MW: 312.077 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34824232
CAS Number: 2918957-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR