Rotors for Centrifuges 5910 Ri and 5920 R
Supplier: Eppendorf
5895130003 5895135005 5895150004 5895120008 5895121004 5895200001 5895201008 5895175007
75874-862EA 2487.02 USD
75874-862 75874-864 75874-866 75875-260 75875-262 76205-036 76205-038 76325-380
Rotors for Centrifuges 5910 Ri and 5920 R
Centrifuges Rotors
Eppendorf rotors for centrifuges 5910 Ri and 5920 R - precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds. Accessorise with versatile buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Experience precision in refrigerated large-capacity centrifugation with Eppendorf rotors designed for centrifuges 5910 Ri and 5920 R. These rotors offer a range of capacities and speeds, providing flexibility for various sample processing needs. Accessorise with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-throughput applications, the Centrifuges 5910 Ri and 5920 R are further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your refrigerated large-capacity centrifugation experience.