Eppendorf® 5810 Benchtop Centrifuge
Supplier: Eppendorf
022625004 022627007 022627066 022628179 022628056 022628078 022627110 022628185 022628090 022628157
53513-808EA 11872.3 USD
53513-808 97007-372 97007-376 89210-982 89210-984 89210-986 89305-170 89305-176 89305-182 75856-496
Eppendorf® 5810 Benchtop Centrifuge
Centrifuges Benchtop Centrifuges
The eppendorf centrifuge 5810 is the perfect day-to-day centrifuge for laboratories with medium-throughput requirements. It has an extremely compact footprint yet still offers the real versatility of being able to take both tube and plate formats.
  • Centrifuge lid with soft-touch lid closure
  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Aerosol-tight Eppendorf
More Product Information