Blue Super-Duty Autoclavable SMS Muff Covers
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MFKCRP600-03 MFKCRP600-04 MFKCRP600-05 MFKCRP600-06 MFKCRP600-07 MFKCRP600-08 MFKCRP600-09 MFKCRP600-12 MFKCRP600-16 MFKCRP600-18
77395-132CS 1567.5 USD
77395-132 77395-134 77395-136 77395-138 77395-140 77395-142 77395-144 77395-146 77395-148 77395-150
Blue Super-Duty Autoclavable SMS Muff Covers
Covers
This 88 g/m² SMS autoclavable muff cover provides an excellent microbial barrier that is ideal for pharmaceutical production applications.
  • Eliminates autoclave tape residue
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
  • Made in USA
Eliminate the need for cutting sheets of sterilization wrapping materials and securing them with autoclave tape and rubber bands. Cost-effective, time-saving approach standardizes the equipment preparation process, minimizes particulate generation, while also ensuring good aseptic technique upon removal after sterilization.
Certificaciones: Certificates of conformance are sent with every shipment.
Envase: Cleanroom packed in sealed, labeled poly-bags with the lot number and date of manufacture. Covers are packed 100 per bag.