WEBCOL™ and CURITY® Alcohol Preps, Covidien
Supplier: Cardinal Health
5110 6818-
82003-856CS 160.28 USD
82003-856 89029-964 82004-802
WEBCOL™ and CURITY® Alcohol Preps, Covidien
Wipes Disposable Wipes and Towels Disinfectant Wipes
These two-ply nonwoven preparatory pads are designed for moderate cleansing and disinfecting.
All pads are saturated with 70% isopropyl alcohol.
WEBCOL™ pads provide maximum absorbency for scrubbing and cleansing. CURITY® pads are made of a lighter weight material and are a more economical choice.
Información para pedidos: Each pad is sealed in an airtight, poly-lined foil pouch. Pouches are packaged in boxes of 200; each case contains 20 boxes.