RCRA Hazardous Waste Containers, Covidien
Supplier: Cardinal Health
8608RC 8612RC 8618RC
89002-258CS 565.24 USD
89002-258 89002-260 89004-412
RCRA Hazardous Waste Containers, Covidien
Bins Waste Bins
Black containers with white sliding-top lids are easily distinguished from sharps, chemo, and other disposal containers
PGII rated containers meet DOT requirements for transport without secondary packaging. RCRA Hazardous Waste labels meet EPA satellite accumulation labeling requirements.
Containers have a leak-resistant gasket and absorbent pad to help contain liquid contents.
Información para pedidos: Toxic, flammable, and tamper evident labels are included in each case to designate contents. For floor carts, contact your Avantor sales representative.