PlateLoc Microplate Heat Seals
Supplier: Agilent Technologies
06644-001 16985-001 17318-001 24210-001 24212-001 24214-001
77463-606EA 2092.21 USD
77463-606 77463-626 77463-628 77463-712 77463-714 77463-716
PlateLoc Microplate Heat Seals
Seals Microplate Seals
Certified for use with the PlateLoc Thermal Microplate Sealer.
  • Choice of aluminum-based or optically clear materials with peelable, pierceable or permanent sealants
  • Seals to match most microplate materials and applications, including polypropylene, polystyrene and COC
Agilent offers a range of PlateLoc microplate heat seals made from aluminum-based or optically clear materials with peelable, pierceable or permanent sealants matching most microplate materials and applications.
Ideal for life science applications that require automated microplateMore Product Information