Diamond® RealSeal™ Bottles, Wide Mouth, Square, HDPE, Nature
Supplier: Globe Scientific
7180060 7180175 7180250 7180500 7181000
77447-992PK 29.62 USD
77447-992 77447-994 77447-996 77447-998 77448-000
Diamond® RealSeal™ Bottles, Wide Mouth, Square, HDPE, Nature
Bottles Plastic Bottles
Diamond RealSeal™ Square plastic laboratory bottles from Globe Scientific meet the requirements of the most demanding applications.
  • Safe for freezer storage down to –100 °C
  • Resistant to most corrosives and suitable for a wide variety of laboratory uses
  • HDPE with PP Closure
These new additions to the Diamond RealSeal offering expand the range with a selection of bottles that store and pack more efficently than standard round bottles. They are available in several material options.
These new bottles are precision molded from virgin resins forMore Product Information