Sulphuric acid 95.0-98.0% (by acidimetry) NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS US
Danger


Management of Change (MOC) category = R

CH20-02 CH20-03
JTCH20-02EA 227.04 USD
JTCH20-02 JTCH20-03
Sulphuric acid 95.0-98.0% (by acidimetry) NF, Multi-Compendial, J.T.Baker®
Sulfuric acid
Formula: H₂SO₄
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Passes test Bulk pharmaceutical chemical
≤ 0.005 % NF - Residue on Ignition
≤ 1 ppm NF - Chloride (Cl)
Passes Test EP/BP - Chloride (Cl)
95.0 - 98.0 % NF - Reducing Substances
≤ 50 ppm EP/BP - Appearance of Solution
≤ 25 ppm EP/BP - Nitrate (NO₃)
95.0 - 100.5 % EP/BP - Iron (Fe)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR