2-Chloro-5-ethoxypyridine ≥95%

Supplier: AOBChem USA

Warning

98002-5G 98002-25G
77497-168EA 152.44 USD
77497-168 77506-056
2-Chloro-5-ethoxypyridine ≥95%
2-Chloro-5-ethoxypyridine

Precaución: For research use only

Formula: C₇H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD09835252
CAS Number: 856851-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR