2-Chloro-3-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

84933-5G 84933-25G
77496-722EA 114.33 USD
77496-722 77505-732
2-Chloro-3-methoxyphenol ≥95%
2-Chloro-3-methoxyphenol

Precaución: For research use only

Formula: C₇H₇ClO₂
MW: 158.5823 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08275690
CAS Number: 72232-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR