5-Bromo-1,3-dimethyl-2-pyridone ≥95%

Supplier: AOBChem USA

Warning

83396-5G 83396-25G
77496-688EA 150.38 USD
77496-688 77505-698
5-Bromo-1,3-dimethyl-2-pyridone ≥95%
5-Bromo-1,3-dimethyl-2-pyridone

Precaución: For research use only

Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD16658881
CAS Number: 51417-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR