3,5-Dichloro-4-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

80836-5G 80836-25G
77496-552EA 121.54 USD
77496-552 77505-596
3,5-Dichloro-4-formylbenzoic acid ≥97%
3,5-Dichloro-4-formylbenzoic acid

Precaución: For research use only

Formula: C₈H₄Cl₂O₃
MW: 219.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24483956
CAS Number: 153203-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR