4'-Methoxy[1,1'-biphenyl]-3-amine ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Amino-4'-methoxybiphenyl hydrochloride, 4'-Methoxy[1,1'-biphenyl]-3-amine
78457-5G
78457-25G
77496-424EA
112.27
USD
77496-424
77505-500
4'-Methoxy[1,1'-biphenyl]-3-amine ≥97%
4'-Methoxy[1,1'-biphenyl]-3-amine
Precaución: For research use only
Formula:
C₁₃H₁₃NO MW: 199.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03424668 CAS Number: 53059-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...