2,3-Difluoro-4-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Difluoro-4-formylbenzotrifluoride

70703-5G 70703-25G
77496-040EA 125.66 USD
77496-040 77505-230
2,3-Difluoro-4-(trifluoromethyl)benzaldehyde ≥95%
2,3-Difluoro-4-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₈H₃F₅O
MW: 210.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236254
CAS Number: 134099-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR