2-Bromo-4-chloro-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

68922-5G 68922-25G
77495-904EA 162.74 USD
77495-904 77505-120
2-Bromo-4-chloro-5-fluorobenzaldehyde ≥97%
2-Bromo-4-chloro-5-fluorobenzaldehyde

Precaución: For research use only

Formula: C₇H₃BrClFO
Storage Temperature: Ambient
MDL Number: MFCD20486524
CAS Number: 1067882-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR