4-Chloro-2,6-difluoroanisole ≥97%

Supplier: AOBChem USA

Warning

64830-5G 64830-25G
77495-622EA 73.13 USD
77495-622 77504-926
4-Chloro-2,6-difluoroanisole ≥97%
4-Chloro-2,6-difluoroanisole

Precaución: For research use only

Formula: C₇H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD13193248
CAS Number: 170572-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR