2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,2,2-Trifluoro-1-(4-(trifluoromethyl)phenyl)ethanone, 4-(Trifluoroacetyl)benzotrifluoride, 2,2,2-Trifluoro-1-[4-(trifluoromethyl)phenyl]ethan-1-one

63299-5G 63299-25G
77495-466EA 60.77 USD
77495-466 77504-800
2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone ≥95%
2,2,2-Trifluoro-4'-(trifluoromethyl)acetophenone

Precaución: For research use only

Formula: C₉H₄F₆O
MW: 242.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061276
CAS Number: 74853-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR