1-(3-(tert-Butyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

63107-5G 63107-25G
77495-456EA 92.7 USD
77495-456 77504-794
1-(3-(tert-Butyl)phenyl)ethanone ≥95%
1-(3-(tert-Butyl)phenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₆O
MW: 176.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113485
CAS Number: 6136-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR