4-Chloro-3,5-difluoroaniline ≥97%

Supplier: AOBChem USA

Warning

60432-5G 60432-25G
77495-312EA 121.54 USD
77495-312 77504-692
4-Chloro-3,5-difluoroaniline ≥97%
4-Chloro-3,5-difluoroaniline

Precaución: For research use only

Formula: C₆H₄ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD09833604
CAS Number: 2613-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR