4-Chloro-3-iodobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

56664-5G 56664-25G
77495-128EA 151.41 USD
77495-128 77504-570
4-Chloro-3-iodobenzaldehyde ≥95%
4-Chloro-3-iodobenzaldehyde

Precaución: For research use only

Formula: C₇H₄ClIO
Storage Temperature: Ambient
MDL Number: MFCD09836681
CAS Number: 276866-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR