1-(6-Fluoropyridin-3-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

54064-5G 54064-25G
77494-986EA 154.5 USD
77494-986 77504-456
1-(6-Fluoropyridin-3-yl)ethanone ≥97%
1-(6-Fluoropyridin-3-yl)ethanone

Precaución: For research use only

Formula: C₇H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD07375076
CAS Number: 84331-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR