2',3'-Difluoro-4'-methylacetophenone ≥97%

Supplier: AOBChem USA

Warning

53217-5G 53217-25G
77494-926EA 86.52 USD
77494-926 77504-408
2',3'-Difluoro-4'-methylacetophenone ≥97%
2',3'-Difluoro-4'-methylacetophenone

Precaución: For research use only

Formula: C₉H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD01631650
CAS Number: 261763-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR