2-Chloro-4,6-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

49220-5G 49220-25G
77494-654EA 109.18 USD
77494-654 77504-226
2-Chloro-4,6-difluorophenol ≥97%
2-Chloro-4,6-difluorophenol

Precaución: For research use only

Formula: C₆H₃ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD11501032
CAS Number: 2267-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR