2,3-Difluoro-4-iodophenol ≥97%

Supplier: AOBChem USA

Warning

48609-5G 48609-25G
77494-600EA 117.42 USD
77494-600 77504-186
2,3-Difluoro-4-iodophenol ≥97%
2,3-Difluoro-4-iodophenol

Precaución: For research use only

Formula: C₆H₃F₂IO
Storage Temperature: Ambient
MDL Number: MFCD08741391
CAS Number: 144292-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR