Methyl 4-bromothiophene-3-carboxylate ≥95%

Supplier: AOBChem USA

Warning

47290-5G 47290-25G
77494-518EA 60.77 USD
77494-518 77504-124
Methyl 4-bromothiophene-3-carboxylate ≥95%
Methyl 4-bromothiophene-3-carboxylate

Precaución: For research use only

Formula: C₆H₅BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD12913867
CAS Number: 78071-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR