3-Fluoro-5-methylbenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (3-Fluoro-5-methylphenyl)methanol

388253-5G 388253-25G
77494-116EA 60.77 USD
77494-116 77503-824
3-Fluoro-5-methylbenzyl alcohol ≥95%
3-Fluoro-5-methylbenzyl alcohol

Precaución: For research use only

Formula: C₈H₉FO
MW: 140.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094314
CAS Number: 518070-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR