4-Ethoxy-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37040-5G 37040-25G
77493-808EA 125.66 USD
77493-808 77503-552
4-Ethoxy-3-(trifluoromethyl)benzaldehyde ≥97%
4-Ethoxy-3-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06247417
CAS Number: 883541-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR