1-(4-Bromophenyl)ethyl methyl ether ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-(1-methoxyethyl)benzene

35497-5G 35497-25G
77493-256EA 131.84 USD
77493-256 77503-042
1-(4-Bromophenyl)ethyl methyl ether ≥95%
1-(4-Bromophenyl)ethyl methyl ether

Precaución: For research use only

Formula: C₉H₁₁BrO
MW: 215.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12027065
CAS Number: 59891-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR