1-Bromo-4-(cyclohexyloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

34991-5G 34991-25G
77493-182EA 138.02 USD
77493-182 77502-982
1-Bromo-4-(cyclohexyloxy)benzene ≥97%
1-Bromo-4-(cyclohexyloxy)benzene

Precaución: For research use only

Formula: C₁₂H₁₅BrO
MW: 255.1509 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11114686
CAS Number: 30752-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR