N-(2,6-Difluoro-3-nitrophenyl)acetamide ≥95%

Supplier: AOBChem USA

Warning

345306-5G 345306-25G
77492-988EA 60.77 USD
77492-988 77502-802
N-(2,6-Difluoro-3-nitrophenyl)acetamide ≥95%
N-(2,6-Difluoro-3-nitrophenyl)acetamide

Precaución: For research use only

Formula: C₈H₆F₂N₂O₃
MW: 216.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174326
CAS Number: 25892-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR