1,3-Difluoro-2,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

33745-5G 33745-25G
77492-736EA 148.32 USD
77492-736 77502-572
1,3-Difluoro-2,4-dimethoxybenzene ≥97%
1,3-Difluoro-2,4-dimethoxybenzene

Precaución: For research use only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09800643
CAS Number: 195136-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR