2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine

331833-5G 331833-25G
77492-648EA 121.54 USD
77492-648 77502-500
2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine ≥95%
2-(2,4-Difluorophenyl)-5-(trifluoromethyl)pyridine

Precaución: For research use only

Formula: C₁₂H₆F₅N
Storage Temperature: Ambient
MDL Number: MFCD28144542
CAS Number: 387827-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR