5-Bromo-2-chloropyridine 1-oxide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-chloropyridine N-oxide

29370-5G 29370-25G
77491-366EA 135.96 USD
77491-366 77501-362
5-Bromo-2-chloropyridine 1-oxide ≥97%
5-Bromo-2-chloropyridine 1-oxide

Precaución: For research use only

Formula: C₅H₃BrClNO
MW: 208.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265108
CAS Number: 889865-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR