Methyl thieno[2,3-b]pyridine-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

28571-5G 28571-25G
77491-198EA 106.09 USD
77491-198 77501-228
Methyl thieno[2,3-b]pyridine-2-carboxylate ≥97%
Methyl thieno[2,3-b]pyridine-2-carboxylate

Precaución: For research use only

Formula: C₉H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD12025885
CAS Number: 154650-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR