3-(Pyridin-3-yl)aniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Pyridin-3-yl)benzenamine

27553-5G 27553-25G
77491-020EA 147.29 USD
77491-020 77501-072
3-(Pyridin-3-yl)aniline ≥97%
3-(Pyridin-3-yl)aniline

Precaución: For research use only

Formula: C₁₁H₁₀N₂
MW: 170.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368549
CAS Number: 57976-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR