1-Methyl-1H-indazole-5-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Methyl-1H-indazole-5-carbaldehyde

27153-5G 27153-25G
77490-958EA 117.42 USD
77490-958 77501-014
1-Methyl-1H-indazole-5-carboxaldehyde ≥97%
1-Methyl-1H-indazole-5-carboxaldehyde

Precaución: For research use only

Formula: C₉H₈N₂O
MW: 160.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109365
CAS Number: 872607-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR