2-Bromoisophthalic acid ≥97%

Supplier: AOBChem USA

Warning

25111-5G 25111-25G
77490-508EA 130.81 USD
77490-508 77500-654
2-Bromoisophthalic acid ≥97%
2-Bromoisophthalic acid

Precaución: For research use only

Formula: C₈H₅BrO₄
Storage Temperature: Ambient
MDL Number: MFCD00099229
CAS Number: 22433-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR